Product Details
- SNP ID
-
rs3099597
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:13495857 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TAGAAATGGTACACATTAAATAATA[A/G]CTTGATTATCAAAATCTCATGAGAT
- Phenotype
-
MIM: 168450
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PTH
PubMed Links
Gene Details
- Gene
- PTH
- Gene Name
- parathyroid hormone
View Full Product Details