Product Details

SNP ID
hCV27460141
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:87116841 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTGATCTTGGCCTGGATTTTATG[C/T]GTCAGTGTGACAGAGAGGGTAAGGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM35A PubMed Links

Gene Details

Gene
FAM35A
Gene Name
family with sequence similarity 35 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019054.2 Intron NP_061927.2

View Full Product Details