Product Details

SNP ID
rs3171603
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:30334337 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCAGTTGCTAATGGGCTGCTGAA[A/G]AAATGAATGTCCTCAAATCGAGACC
Phenotype
MIM: 605595 MIM: 610020
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SF3A1 PubMed Links

Gene Details

Gene
SF3A1
Gene Name
splicing factor 3a subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005877.5 2794 UTR 3 NP_005868.1
Gene
TBC1D10A
Gene Name
TBC1 domain family member 10A
There are no transcripts associated with this gene.

View Full Product Details