Product Details

SNP ID
rs3169716
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:44248453 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCATCAAGGGGAAGGGTGCTAGCA[A/G]AGTGAATAATTTTTGTTTTTCACTT
Phenotype
MIM: 109270
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC4A1 PubMed Links

Gene Details

Gene
SLC4A1
Gene Name
solute carrier family 4 member 1 (Diego blood group)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000342.3 4890 UTR 3 NP_000333.1
XM_005257593.4 4890 UTR 3 XP_005257650.1
XM_011525129.1 4890 UTR 3 XP_011523431.1
XM_011525130.1 4890 Intron XP_011523432.1

View Full Product Details