Product Details

SNP ID
rs3176435
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:86096 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAGGGCCAGCCCAGGCCCTCGTG[A/C]CCCAGATGGTCAGGACCAGGTCACA
Phenotype
MIM: 156565 MIM: 600928 MIM: 614403
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MPG PubMed Links

Gene Details

Gene
MPG
Gene Name
N-methylpurine DNA glycosylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001015052.2 2643 Intron NP_001015052.1
NM_001015054.2 2643 Intron NP_001015054.1
NM_002434.3 2643 Intron NP_002425.2
Gene
NPRL3
Gene Name
NPR3 like, GATOR1 complex subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039476.2 2643 UTR 3 NP_001034565.1
NM_001077350.2 2643 UTR 3 NP_001070818.1
NM_001243247.1 2643 UTR 3 NP_001230176.1
NM_001243248.1 2643 UTR 3 NP_001230177.1
NM_001243249.1 2643 UTR 3 NP_001230178.1
Gene
RHBDF1
Gene Name
rhomboid 5 homolog 1
There are no transcripts associated with this gene.

View Full Product Details