Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_024589.2 | 1623 | UTR 3 | NP_078865.1 | ||
XM_006720947.3 | 1623 | UTR 3 | XP_006721010.1 | ||
XM_006720948.3 | 1623 | UTR 3 | XP_006721011.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001253790.1 | 1623 | Intron | NP_001240719.1 | ||
NM_001253791.1 | 1623 | Intron | NP_001240720.1 | ||
NM_001253793.1 | 1623 | Intron | NP_001240722.1 | ||
NM_001253794.1 | 1623 | Intron | NP_001240723.1 | ||
XM_011522499.2 | 1623 | Intron | XP_011520801.1 | ||
XM_011522500.2 | 1623 | Intron | XP_011520802.1 | ||
XM_011522501.2 | 1623 | Intron | XP_011520803.1 |