Product Details

SNP ID
rs3813546
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:76153618 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGAGGGTAGGAGAGGGGCTTAAAA[A/G]TTGTTAATTATTCAAAAATATATGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPATCH2L PubMed Links

Gene Details

Gene
GPATCH2L
Gene Name
G-patch domain containing 2 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322026.1 Intron NP_001308955.1
NM_001322027.1 Intron NP_001308956.1
NM_001322028.1 Intron NP_001308957.1
NM_001322029.1 Intron NP_001308958.1
NM_001322030.1 Intron NP_001308959.1
NM_001322031.1 Intron NP_001308960.1
NM_001322032.1 Intron NP_001308961.1
NM_017926.3 Intron NP_060396.2
NM_017972.3 Intron NP_060442.2
XM_006720191.2 Intron XP_006720254.1
XM_017021427.1 Intron XP_016876916.1
XM_017021428.1 Intron XP_016876917.1
XM_017021429.1 Intron XP_016876918.1
XM_017021430.1 Intron XP_016876919.1
XM_017021431.1 Intron XP_016876920.1
XM_017021432.1 Intron XP_016876921.1
XM_017021433.1 Intron XP_016876922.1

View Full Product Details