Product Details
- SNP ID
-
rs3744752
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:1004394 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCAAAAAGGGCCTCTCACCGCGCAC[A/G]CGCTGCAGCGTTAGGGCCGGCAAAC
- Phenotype
-
MIM: 600365
MIM: 607251
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ABR
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs143173545] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ABR
- Gene Name
- active BCR-related
- Gene
- TIMM22
- Gene Name
- translocase of inner mitochondrial membrane 22 homolog (yeast)
There are no transcripts associated with this gene.
View Full Product Details