Product Details

SNP ID
rs3763752
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:121742635 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAAAGAAAGTGAGGGAAGGAGGGG[C/G]CTGGCCTCTGCTCTGAGTTACTAAG
Phenotype
MIM: 607103
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ATE1 PubMed Links

Gene Details

Gene
ATE1
Gene Name
arginyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001976.2 2496 UTR 3 NP_001001976.1
NM_001288734.1 2496 UTR 3 NP_001275663.1
NM_001288735.1 2496 UTR 3 NP_001275664.1
NM_001288736.1 2496 UTR 3 NP_001275665.1
NM_007041.3 2496 UTR 3 NP_008972.2
XM_005269458.2 2496 UTR 3 XP_005269515.1
XM_005269459.3 2496 UTR 3 XP_005269516.1
XM_017015597.1 2496 UTR 3 XP_016871086.1
XM_017015598.1 2496 UTR 3 XP_016871087.1
XM_017015599.1 2496 UTR 3 XP_016871088.1
XM_017015600.1 2496 UTR 3 XP_016871089.1
XM_017015601.1 2496 Intron XP_016871090.1
XM_017015602.1 2496 UTR 3 XP_016871091.1
XM_017015603.1 2496 Intron XP_016871092.1
XM_017015604.1 2496 UTR 3 XP_016871093.1

View Full Product Details