Product Details

SNP ID
rs3740702
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:45648845 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGACCCGGCTGAGGCCCTCTCGTA[C/T]CCATTCAGAATGTCCTGCAATCACA
Phenotype
MIM: 603797
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHST1 PubMed Links

Gene Details

Gene
CHST1
Gene Name
carbohydrate sulfotransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003654.5 2784 UTR 3 NP_003645.1
XM_006718356.3 2784 Intron XP_006718419.1
XM_017018459.1 2784 Intron XP_016873948.1

View Full Product Details