Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001144070.1 | Intron | NP_001137542.1 | |||
NM_003786.3 | Intron | NP_003777.2 | |||
XM_005257763.3 | Intron | XP_005257820.1 | |||
XM_011525422.2 | Intron | XP_011523724.1 | |||
XM_011525423.1 | Intron | XP_011523725.1 | |||
XM_011525424.2 | Intron | XP_011523726.1 | |||
XM_011525425.2 | Intron | XP_011523727.1 | |||
XM_017025265.1 | Intron | XP_016880754.1 | |||
XM_017025266.1 | Intron | XP_016880755.1 |