Product Details

SNP ID
rs3785931
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:49500756 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAACACCAAGGTTCCACCTCCTCTG[C/T]TGCCTGTGCCTGTCCTGCTGGTCTC
Phenotype
MIM: 162010
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100288866 PubMed Links

Gene Details

Gene
LOC100288866
Gene Name
uncharacterized LOC100288866
There are no transcripts associated with this gene.

Gene
NGFR
Gene Name
nerve growth factor receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002507.3 Intron NP_002498.1

View Full Product Details