Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_014705.3 | 7910 | UTR 3 | NP_055520.3 | ||
XM_006716188.2 | 7910 | Intron | XP_006716251.1 | ||
XM_006716189.2 | 7910 | Intron | XP_006716252.1 | ||
XM_017012819.1 | 7910 | Intron | XP_016868308.1 | ||
XM_017012820.1 | 7910 | Intron | XP_016868309.1 | ||
XM_017012821.1 | 7910 | UTR 3 | XP_016868310.1 | ||
XM_017012822.1 | 7910 | Intron | XP_016868311.1 | ||
XM_017012823.1 | 7910 | Intron | XP_016868312.1 | ||
XM_017012824.1 | 7910 | UTR 3 | XP_016868313.1 | ||
XM_017012825.1 | 7910 | UTR 3 | XP_016868314.1 |