Product Details

SNP ID
rs3809197
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:96489640 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCGCAGGGCTCCCCCTCGAGCTCT[C/T]CGTCAGACGACTCTACCACCTCGGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CFAP54 PubMed Links

Gene Details

Gene
CFAP54
Gene Name
cilia and flagella associated protein 54
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001306084.1 75 Missense Mutation CCG,TCG P11S NP_001293013.1
XM_011539069.2 75 Missense Mutation CCG,TCG P11S XP_011537371.1
XM_011539070.2 75 Missense Mutation CCG,TCG P11S XP_011537372.1
XM_011539071.2 75 Missense Mutation CCG,TCG P11S XP_011537373.1
XM_011539072.2 75 Missense Mutation CCG,TCG P11S XP_011537374.1
XM_011539073.2 75 Missense Mutation CCG,TCG P11S XP_011537375.1
XM_011539074.2 75 Missense Mutation CCG,TCG P11S XP_011537376.1
XM_011539076.2 75 Missense Mutation CCG,TCG P11S XP_011537378.1
XM_011539077.2 75 Intron XP_011537379.1
XM_011539078.2 75 Missense Mutation CCG,TCG P11S XP_011537380.1
XM_011539079.1 75 Intron XP_011537381.1
XM_011539081.2 75 Missense Mutation CCG,TCG P11S XP_011537383.1
XM_011539083.2 75 Missense Mutation CCG,TCG P11S XP_011537385.1
XM_011539084.2 75 Missense Mutation CCG,TCG P11S XP_011537386.1
XM_011539085.1 75 Intron XP_011537387.1
XM_017018866.1 75 Missense Mutation CCG,TCG P11S XP_016874355.1
XM_017018867.1 75 Intron XP_016874356.1

View Full Product Details