Product Details

SNP ID
rs4073174
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:1069402 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCGTGGCCTGTGCGGGCGTCGCC[C/A]TGGTGACCGGCGGCCCCTCCCCCCA
Phenotype
Polymorphism
C/A, Transversion substitution
Allele Nomenclature
Literature Links
LOC100288175 PubMed Links

Gene Details

Gene
LOC100288175
Gene Name
uncharacterized LOC100288175
There are no transcripts associated with this gene.

Gene
LOC105378948
Gene Name
uncharacterized LOC105378948
There are no transcripts associated with this gene.

Gene
RNF223
Gene Name
ring finger protein 223
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001205252.1 3466 Intron NP_001192181.1
XM_017001286.1 3466 UTR 3 XP_016856775.1
XM_017001287.1 3466 Intron XP_016856776.1

View Full Product Details