Product Details

SNP ID
rs3813020
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30922754 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCTGCAGCCGCCATCTTTGCCGGT[G/A]GCCCCGCCCCATCCCCTTCCATTGT
Phenotype
MIM: 609085
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
FBXL19 PubMed Links

Gene Details

Gene
FBXL19
Gene Name
F-box and leucine rich repeat protein 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099784.2 Intron NP_001093254.2
NM_001282351.1 Intron NP_001269280.1
Gene
FBXL19-AS1
Gene Name
FBXL19 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details