Product Details

SNP ID
hCV27531084
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:32064171 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGCCGGTGCCGGGCGGCGAGGCC[G/T]AGCGCGTCCGAGTCTTCCACAAACA
Phenotype
MIM: 604277
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SPAST PubMed Links

Gene Details

Gene
SPAST
Gene Name
spastin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014946.3 561 Nonsense Mutation GAG,TAG E114* NP_055761.2
NM_199436.1 561 Nonsense Mutation GAG,TAG E114* NP_955468.1

View Full Product Details