Product Details
- SNP ID
-
rs121964985
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:49417892 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCCCCCTCACACATCAACCCCACA[T/C]GCCTCCGCTGCACCCTGCCCTTCAG
- Phenotype
-
MIM: 238310
MIM: 611516
MIM: 165390
MIM: 600690
- Polymorphism
- T/C, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
AMT
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11715915] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AMT
- Gene Name
- aminomethyltransferase
- Gene
- NICN1
- Gene Name
- nicolin 1
There are no transcripts associated with this gene.
- Gene
- RHOA
- Gene Name
- ras homolog family member A
There are no transcripts associated with this gene.
- Gene
- TCTA
- Gene Name
- T-cell leukemia translocation altered
There are no transcripts associated with this gene.
View Full Product Details