Product Details

SNP ID
rs797044527
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:138946073 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCTGCGGCTGCCGCCATCTGGCAG[A/G]AGGCATAGGGCATGGGTGAGGGAGG
Phenotype
MIM: 605597
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FOXL2 PubMed Links

Gene Details

Gene
FOXL2
Gene Name
forkhead box L2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_023067.3 1068 Missense Mutation TCC,TTC S217F NP_075555.1
Gene
FOXL2NB
Gene Name
FOXL2 neighbor
There are no transcripts associated with this gene.

Gene
LINC01391
Gene Name
long intergenic non-protein coding RNA 1391
There are no transcripts associated with this gene.

View Full Product Details