Product Details

SNP ID
rs104893837
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:67740682 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCGTCATTTTTAGAGTCTTCAGC[C/T]GTGCTCTTGGTATATTGTTCTTGGA
Phenotype
MIM: 138850
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GNRHR PubMed Links

Gene Details

Gene
GNRHR
Gene Name
gonadotropin releasing hormone receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000406.2 2536 Missense Mutation CAG,CGG Q262R NP_000397.1
NM_001012763.1 2536 Silent Mutation ACA,ACG T219T NP_001012781.1

View Full Product Details