Product Details

SNP ID
rs28933074
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:67740616 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACCAATACCAAATTCCTAGGACA[C/T]AGTAGGGAGTCCAGCAGACAGTAAA
Phenotype
MIM: 138850
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GNRHR PubMed Links

Gene Details

Gene
GNRHR
Gene Name
gonadotropin releasing hormone receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000406.2 2602 Missense Mutation TAT,TGT Y284C NP_000397.1
NM_001012763.1 2602 Silent Mutation CTA,CTG L241L NP_001012781.1

View Full Product Details