Product Details

SNP ID
rs112354911
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:95202020 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGGCCTTACCCCGCCCTCTGGAA[C/G]GTTAGTGCCAAAGGAGAAGAGGACA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ACSM6 PubMed Links

Gene Details

Gene
ACSM6
Gene Name
acyl-CoA synthetase medium-chain family member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207321.2 353 Missense Mutation AAC,AAG N76K NP_997204.2

View Full Product Details