Product Details

SNP ID
rs58284890
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:43629979 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAACCAGCTTCCTCCCCGAAGGCT[A/G]CAAGCATACCCAAAGCGAGTATTCC
Phenotype
MIM: 607434 MIM: 603968
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GTPBP2 PubMed Links

Gene Details

Gene
GTPBP2
Gene Name
GTP binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286216.1 Intron NP_001273145.1
NM_019096.4 Intron NP_061969.3
XM_017010976.1 Intron XP_016866465.1
Gene
MAD2L1BP
Gene Name
MAD2L1 binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003690.1 Intron NP_001003690.1
NM_014628.2 Intron NP_055443.1
Gene
POLH
Gene Name
DNA polymerase eta
There are no transcripts associated with this gene.

View Full Product Details