Product Details

SNP ID
rs77682947
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:49550035 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTACCGCCTTTTTTTCATATAATG[C/T]AAGCAAAGAGTTGCCAGTTGCAACA
Phenotype
MIM: 604020
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BSN PubMed Links

Gene Details

Gene
BSN
Gene Name
bassoon presynaptic cytomatrix protein
There are no transcripts associated with this gene.

Gene
BSN-AS2
Gene Name
BSN antisense RNA 2 (head to head)
There are no transcripts associated with this gene.

View Full Product Details