Product Details

SNP ID
rs1055674717
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:122301875 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGGGGATGCAGAGGCTTCTGGTC[C/G]GCCCTGAAGTGTGGGGCCCAACATG
Phenotype
MIM: 613301
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FEZF1 PubMed Links

Gene Details

Gene
FEZF1
Gene Name
FEZ family zinc finger 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024613.3 1624 UTR 3 NP_001019784.2
NM_001160264.2 1624 UTR 3 NP_001153736.1
Gene
FEZF1-AS1
Gene Name
FEZF1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details