Product Details

SNP ID
rs75576635
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:54308429 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAGTATACTCATTCAAGTAAAGG[A/G]TCAGTTTCAGGGTTCAGGCTGAAGT
Phenotype
MIM: 606749
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TINAG PubMed Links

Gene Details

Gene
TINAG
Gene Name
tubulointerstitial nephritis antigen
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014464.3 25 UTR 5 NP_055279.3
XM_006715062.2 25 Intron XP_006715125.1
XM_011514497.2 25 UTR 5 XP_011512799.1
XM_017010745.1 25 Intron XP_016866234.1
XM_017010746.1 25 UTR 5 XP_016866235.1
XM_017010747.1 25 UTR 5 XP_016866236.1
XM_017010748.1 25 Intron XP_016866237.1

View Full Product Details