Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001370.1 | 42 | Intron | NP_001361.1 | ||
XM_006711956.2 | 42 | Intron | XP_006712019.1 | ||
XM_011532649.2 | 42 | UTR 5 | XP_011530951.1 | ||
XM_011532650.2 | 42 | UTR 5 | XP_011530952.1 | ||
XM_011532652.1 | 42 | Intron | XP_011530954.1 | ||
XM_011532653.1 | 42 | Intron | XP_011530955.1 | ||
XM_011532654.1 | 42 | Intron | XP_011530956.1 | ||
XM_011532655.2 | 42 | Intron | XP_011530957.1 | ||
XM_011532657.1 | 42 | Intron | XP_011530959.1 | ||
XM_011532660.1 | 42 | Intron | XP_011530962.1 | ||
XM_017003521.1 | 42 | Intron | XP_016859010.1 | ||
XM_017003522.1 | 42 | Intron | XP_016859011.1 | ||
XM_017003523.1 | 42 | UTR 5 | XP_016859012.1 | ||
XM_017003524.1 | 42 | Intron | XP_016859013.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_003849.3 | 42 | Intron | NP_003840.2 |