Product Details

SNP ID
rs41309506
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:101348375 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCCCAGAGAAGCGGAGTCACCCT[C/T]CTCCCATCCGGCTAGGCCACAGGAA
Phenotype
MIM: 300300 MIM: 300356
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BTK PubMed Links

Gene Details

Gene
BTK
Gene Name
Bruton tyrosine kinase
There are no transcripts associated with this gene.

Gene
TIMM8A
Gene Name
translocase of inner mitochondrial membrane 8 homolog A (yeast)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145951.1 215 UTR 3 NP_001139423.1
NM_004085.3 215 Intron NP_004076.1

View Full Product Details