Product Details

SNP ID
rs4796424
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7484654 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCCTCCGGCTTTTTCCTCCCAAC[C/T]CGGGGAGGTCCTTCCCGGTGGCCGC
Phenotype
MIM: 180660 MIM: 612308
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
POLR2A PubMed Links

Gene Details

Gene
POLR2A
Gene Name
RNA polymerase II subunit A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000937.4 276 UTR 5 NP_000928.1
Gene
SLC35G6
Gene Name
solute carrier family 35 member G6
There are no transcripts associated with this gene.

Gene
ZBTB4
Gene Name
zinc finger and BTB domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128833.1 276 Intron NP_001122305.1
NM_020899.3 276 Intron NP_065950.2
XM_006721563.3 276 Intron XP_006721626.1
XM_006721564.2 276 Intron XP_006721627.1
XM_011523972.2 276 Intron XP_011522274.1

View Full Product Details