Product Details

SNP ID
rs117419466
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:42562647 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGGCCGTATTCCGGTCGGGTCTC[C/T]TGGTGCTGACGACGCCGCTGGCCTC
Phenotype
MIM: 609855 MIM: 109684 MIM: 602976 MIM: 608665
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COASY PubMed Links

Gene Details

Gene
COASY
Gene Name
Coenzyme A synthase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042529.2 378 Silent Mutation CTG,TTG L9L NP_001035994.1
NM_001042532.3 378 Silent Mutation CTG,TTG L38L NP_001035997.2
NM_025233.6 378 Silent Mutation CTG,TTG L9L NP_079509.5
XM_006722116.3 378 Silent Mutation CTG,TTG L38L XP_006722179.1
XM_011525300.1 378 Silent Mutation CTG,TTG L9L XP_011523602.1
XM_017025167.1 378 Silent Mutation CTG,TTG L9L XP_016880656.1
XM_017025168.1 378 Intron XP_016880657.1
Gene
HSD17B1
Gene Name
hydroxysteroid 17-beta dehydrogenase 1
There are no transcripts associated with this gene.

Gene
MLX
Gene Name
MLX, MAX dimerization protein
There are no transcripts associated with this gene.

Gene
PSMC3IP
Gene Name
PSMC3 interacting protein
There are no transcripts associated with this gene.

View Full Product Details