Product Details

SNP ID
rs138368432
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:158521909 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATAGCACACAACCACTGTGAGCT[A/G]TATGCGATTCAGAAACCAAGACCAA
Phenotype
MIM: 606654
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RXFP1 PubMed Links

Gene Details

Gene
RXFP1
Gene Name
relaxin/insulin like family peptide receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001253727.1 165 UTR 5 NP_001240656.1
NM_001253728.1 165 UTR 5 NP_001240657.1
NM_001253729.1 165 UTR 5 NP_001240658.1
NM_001253730.1 165 UTR 5 NP_001240659.1
NM_001253732.1 165 UTR 5 NP_001240661.1
NM_001253733.1 165 UTR 5 NP_001240662.1
NM_021634.3 165 UTR 5 NP_067647.2
XM_011532174.1 165 UTR 5 XP_011530476.1
XM_011532175.1 165 UTR 5 XP_011530477.1
XM_011532176.1 165 UTR 5 XP_011530478.1
XM_011532179.2 165 UTR 5 XP_011530481.1
XM_017008517.1 165 UTR 5 XP_016864006.1
XM_017008518.1 165 UTR 5 XP_016864007.1
XM_017008519.1 165 UTR 5 XP_016864008.1
XM_017008520.1 165 UTR 5 XP_016864009.1
XM_017008521.1 165 UTR 5 XP_016864010.1
XM_017008522.1 165 UTR 5 XP_016864011.1
XM_017008523.1 165 UTR 5 XP_016864012.1
XM_017008524.1 165 UTR 5 XP_016864013.1
XM_017008525.1 165 UTR 5 XP_016864014.1
XM_017008526.1 165 Intron XP_016864015.1

View Full Product Details