Product Details

SNP ID
rs60921765
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:31380691 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTTACTCAATTATTATTATAAGC[A/G]TTATTCAGGAAAGAACTGCTTATTA
Phenotype
MIM: 607892
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DSG1-AS1 PubMed Links

Gene Details

Gene
DSG1-AS1
Gene Name
DSG1 antisense RNA 1
There are no transcripts associated with this gene.

Gene
DSG4
Gene Name
desmoglein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134453.2 Intron NP_001127925.1
NM_177986.4 Intron NP_817123.1
XM_011525827.1 Intron XP_011524129.1

View Full Product Details