Product Details

SNP ID
rs72882720
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:27343108 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGACCTGCCCGCCAAGCTGCCTC[C/T]GCCGGTCTCAGCCCCACCTGGAGCC
Phenotype
MIM: 608042
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SYTL1 PubMed Links

Gene Details

Gene
SYTL1
Gene Name
synaptotagmin like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193308.1 Intron NP_001180237.1
NM_032872.2 Intron NP_116261.1
XM_005246022.3 Intron XP_005246079.1
XM_006710990.1 Intron XP_006711053.1
XM_006710991.1 Intron XP_006711054.1
Gene
TMEM222
Gene Name
transmembrane protein 222
There are no transcripts associated with this gene.

View Full Product Details