Product Details

SNP ID
rs144904252
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:40180976 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGACTGGCTGAGCCAGGCCCAGAC[A/G]GGGGTGTGGAGGGGAAAGGGGTGGT
Phenotype
MIM: 606504
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CASC3 PubMed Links

Gene Details

Gene
CASC3
Gene Name
cancer susceptibility candidate 3
There are no transcripts associated with this gene.

Gene
RAPGEFL1
Gene Name
Rap guanine nucleotide exchange factor like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303533.1 1510 Intron NP_001290462.1
NM_001303534.1 1510 Intron NP_001290463.1
NM_016339.4 1510 Intron NP_057423.1
XM_006721932.3 1510 Intron XP_006721995.1
XM_006721935.3 1510 Intron XP_006721998.1
XM_006721937.3 1510 UTR 5 XP_006722000.1
XM_006721938.3 1510 Intron XP_006722001.1

View Full Product Details