Product Details

SNP ID
rs9939768
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67185204 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGCCCCGTCCGTTCACACACACTTT[C/G]CAGGAGCTCAGGGCTCGACAGCCAT
Phenotype
MIM: 600659 MIM: 614117 MIM: 605235
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
E2F4 PubMed Links

Gene Details

Gene
E2F4
Gene Name
E2F transcription factor 4
There are no transcripts associated with this gene.

Gene
EXOC3L1
Gene Name
exocyst complex component 3 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178516.3 1922 Missense Mutation CAA,GAA Q561E NP_848611.2
Gene
KIAA0895L
Gene Name
KIAA0895 like
There are no transcripts associated with this gene.

Gene
NOL3
Gene Name
nucleolar protein 3
There are no transcripts associated with this gene.

View Full Product Details