Product Details
- SNP ID
-
rs9261536
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:30163572 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCTCTCTTTCTCTCTCTCTGTCTCT[C/T]AGCCTTGCAGCCGTTTCCCTCTGCG
- Phenotype
-
MIM: 605701
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
TRIM10
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs546780690] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TRIM10
- Gene Name
- tripartite motif containing 10
- Gene
- TRIM15
- Gene Name
- tripartite motif containing 15
View Full Product Details