Product Details

SNP ID
rs1055169971
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:37692422 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCATTGGGCGGAAGACCCTGGGC[A/C]GGCAGGAACTTGCTTGTGATTGGAT
Phenotype
MIM: 614799 MIM: 609977
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C1orf109 PubMed Links

Gene Details

Gene
C1orf109
Gene Name
chromosome 1 open reading frame 109
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303030.1 22 Intron NP_001289959.1
NM_001303031.1 22 Intron NP_001289960.1
NM_017850.2 22 Intron NP_060320.1
Gene
CDCA8
Gene Name
cell division cycle associated 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256875.1 22 UTR 5 NP_001243804.1
NM_018101.3 22 UTR 5 NP_060571.1

View Full Product Details