Product Details

SNP ID
hCV27860956
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:177431749 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTCCTCAGTTGGCTGATTTTTTT[A/C]GGAATTATAAACCATGAAACTAAAC
Phenotype
MIM: 613228
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
AGA PubMed Links

Gene Details

Gene
AGA
Gene Name
aspartylglucosaminidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000027.3 1098 Nonsense Mutation GAA,TAA E334* NP_000018.2
NM_001171988.1 1098 Nonsense Mutation GAA,TAA E324* NP_001165459.1

View Full Product Details