Product Details

SNP ID
rs4715326
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:52792754 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGGAGACCTTGGGGCACTGAAGGC[C/T]TGGAGAAGACTGGGGCAAAACTTGG
Phenotype
MIM: 138359
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GSTA1 PubMed Links

Gene Details

Gene
GSTA1
Gene Name
glutathione S-transferase alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319059.1 720 Intron NP_001305988.1
NM_145740.4 720 Intron NP_665683.1
XM_005249034.3 720 Missense Mutation XP_005249091.1

View Full Product Details