Product Details

SNP ID
rs4842973
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:84570227 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCGCCCGGGCTCAAACAACAGCC[A/G]GAGACGCCCTGCCACGCCCCGTCGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00933 PubMed Links

Gene Details

Gene
LINC00933
Gene Name
long intergenic non-protein coding RNA 933
There are no transcripts associated with this gene.

Gene
UBE2Q2P1
Gene Name
ubiquitin conjugating enzyme E2 Q2 pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details