Product Details

SNP ID
rs4290088
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:198647973 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATACCTTGCTACAGATAATTATTGG[G/T]GTTATCAATGTTTTATAAATTATAA
Phenotype
MIM: 151460
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PTPRC PubMed Links

Gene Details

Gene
PTPRC
Gene Name
protein tyrosine phosphatase, receptor type C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267798.1 Intron NP_001254727.1
NM_002838.4 Intron NP_002829.3
NM_080921.3 Intron NP_563578.2

View Full Product Details