Product Details

SNP ID
rs3219145
Assay Type
Functionally tested
NCBI dbSNP Submissions
34
Location
Chr.1:226363128 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GACACTGTGCTTGCCCTTGGGTAAC[C/T]TGCTGATATGTGAAGCGTGCTTCAG
Phenotype
MIM: 173870
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PARP1 PubMed Links

Gene Details

Gene
PARP1
Gene Name
poly(ADP-ribose) polymerase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001618.3 2990 Missense Mutation NP_001609.2

View Full Product Details