Product Details

SNP ID
rs4900699
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:37201764 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTGCTCAGTATTGCCCAAAATTT[A/T]ACTTTAATACCATTCAGCCCACCAT
Phenotype
MIM: 606850
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MIPOL1 PubMed Links

Gene Details

Gene
MIPOL1
Gene Name
mirror-image polydactyly 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195296.1 Intron NP_001182225.1
NM_001195297.1 Intron NP_001182226.1
NM_138731.6 Intron NP_620059.1

View Full Product Details