Product Details

SNP ID
hCV27939851
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:75637032 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCCCGCTGGAGGCCGGGGTCCAC[A/C]CGGCATGAGTCTGCAGCTCTCCCAC
Phenotype
MIM: 603781
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RECQL5 PubMed Links

Gene Details

Gene
RECQL5
Gene Name
RecQ like helicase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003715.3 Intron NP_001003715.1
NM_001003716.3 Intron NP_001003716.1
NM_004259.6 Intron NP_004250.4
Gene
SMIM5
Gene Name
small integral membrane protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001162995.2 Intron NP_001156467.1
Gene
SMIM6
Gene Name
small integral membrane protein 6
There are no transcripts associated with this gene.

View Full Product Details