Product Details

SNP ID
rs4802076
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:40359410 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCAGAGCATAAGTACCAACCTGGA[A/T]GGGGCGCGCTGGGAGCCCCATGGAG
Phenotype
MIM: 615698
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PLD3 PubMed Links

Gene Details

Gene
PLD3
Gene Name
phospholipase D family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031696.3 Intron NP_001026866.1
NM_001291311.1 Intron NP_001278240.1
NM_012268.3 Intron NP_036400.2
XM_005258704.1 Intron XP_005258761.1
XM_005258707.4 Intron XP_005258764.1
XM_005258708.3 Intron XP_005258765.1
XM_005258709.4 Intron XP_005258766.1
XM_005258710.4 Intron XP_005258767.1
XM_006723122.1 Intron XP_006723185.1
XM_011526692.1 Intron XP_011524994.1
XM_011526693.1 Intron XP_011524995.1
XM_017026546.1 Intron XP_016882035.1
XM_017026547.1 Intron XP_016882036.1
XM_017026548.1 Intron XP_016882037.1
XM_017026549.1 Intron XP_016882038.1

View Full Product Details