Product Details
- SNP ID
-
rs4867319
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:31200250 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGAGAATAAATTATCTTATCCTGC[C/T]TTCAGAGGGATCTTTATGAGGAAAG
- Phenotype
-
MIM: 603007
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CDH6
PubMed Links
Gene Details
- Gene
- CDH6
- Gene Name
- cadherin 6
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004932.3 |
|
Intron |
|
|
NP_004923.1 |
View Full Product Details