Product Details
- SNP ID
-
rs4927925
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:129005951 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTTTGTTTATTTCTAAAATCACAA[C/G]TAGTCCTCTATAGCAGAATGCTACA
- Phenotype
-
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
EFCC1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76149190] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- EFCC1
- Gene Name
- EF-hand and coiled-coil domain containing 1
- Gene
- KIAA1257
- Gene Name
- KIAA1257
View Full Product Details