Product Details

SNP ID
rs4715223
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:13362995 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTAAAGACACCTGTTGGACTTCA[C/G]TGAGCCTGTGAACAAAAGCTACGTA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GFOD1 PubMed Links

Gene Details

Gene
GFOD1
Gene Name
glucose-fructose oxidoreductase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242628.1 Intron NP_001229557.1
NM_001242629.1 Intron NP_001229558.1
NM_001242630.1 Intron NP_001229559.1
NM_018988.3 Intron NP_061861.1

View Full Product Details