Product Details

SNP ID
rs4419124
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:48548545 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTGTGTGGGTCACATATTCAGCA[C/G]ATTGGGTAATAAAGAGCATGCGTTG
Phenotype
MIM: 613178
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CTIF PubMed Links

Gene Details

Gene
CTIF
Gene Name
cap binding complex dependent translation initiation factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142397.1 Intron NP_001135869.1
NM_014772.2 Intron NP_055587.1

View Full Product Details