Product Details

SNP ID
rs4293118
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:22201195 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTCATTTGCAGCCACAGTGAAAA[A/C]CTTGGCTCCCTTGTCAGCTCGCTCA
Phenotype
MIM: 608662
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ANO5 PubMed Links

Gene Details

Gene
ANO5
Gene Name
anoctamin 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142649.1 Intron NP_001136121.1
NM_213599.2 Intron NP_998764.1
XM_005252820.3 Intron XP_005252877.2
XM_005252821.3 Intron XP_005252878.2
XM_005252822.4 Intron XP_005252879.1
XM_011519949.2 Intron XP_011518251.1

View Full Product Details